Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115612.3 | NP_191311.1 | 11 - 12 | 1890 | 106 | |
NM_180384.1 | NP_850715.1 | 11 - 12 | 1877 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |