Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP76C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115968.2 | NP_191663.1 | 1 - 2 | 374 | 83 | |
NM_180400.1 | NP_850731.1 | 1 - 2 | 374 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |