Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333396.1 | NP_001320455.1 | 7 - 8 | 1376 | 134 | |
NM_001333397.1 | NP_001320454.1 | 11 - 12 | 2057 | 134 | ||
NM_103851.3 | NP_175386.2 | 11 - 12 | 2086 | 134 | ||
NM_180630.2 | NP_850961.1 | 11 - 12 | 2199 | 134 | ||
NM_180631.2 | NP_850962.1 | 11 - 12 | 2199 | 134 | ||
GenBank mRNA | AY074305.1 | - | 11 - 12 | 2086 | 134 | |
BT004376.1 | - | 11 - 12 | 2041 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |