Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342767.1 | NP_001330089.1 | 18 - 19 | 2112 | 92 | |
NM_120518.5 | NP_196056.2 | 18 - 19 | 2125 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |