Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120544.3 | NP_196082.2 | 7 - 8 | 1638 | 101 | |
NM_203002.4 | NP_974731.1 | 8 - 9 | 1427 | 101 | ||
GenBank mRNA | AK119095.1 | - | 7 - 8 | 1638 | 101 | |
BT026507.1 | - | 6 - 7 | 829 | 101 | ||
DQ017966.1 | - | 8 - 9 | 1246 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |