Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120599.4 | NP_196136.1 | 9 - 10 | 1816 | 73 | |
GenBank mRNA | BT002335.1 | - | 8 - 9 | 1458 | 73 | |
KJ138704.1 | - | 8 - 9 | 1458 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |