Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120769.2 | NP_196304.1 | 1 - 2 | 560 | 68 | |
GenBank mRNA | AF229249.1 | - | 1 - 2 | 537 | 68 | |
AY133567.1 | - | 1 - 2 | 537 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |