Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120778.4 | NP_196313.1 | 2 - 3 | 448 | 80 | |
NM_203016.2 | NP_974745.1 | 2 - 3 | 432 | 80 | ||
GenBank mRNA | AY058830.1 | - | 2 - 3 | 377 | 80 | |
AY143864.1 | - | 1 - 2 | 67 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |