Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125718.1 | NP_001119190.1 | 7 - 8 | 1512 | 89 | |
NM_001203328.2 | NP_001190257.1 | 8 - 9 | 1459 | 89 | ||
NM_001203329.1 | NP_001190258.1 | 8 - 9 | 1459 | 89 | ||
NM_001203330.2 | NP_001190259.1 | 8 - 9 | 1471 | 89 | ||
NM_001342983.1 | NP_001318507.1 | 8 - 9 | 1295 | 89 | ||
NM_001342984.1 | NP_001318508.1 | 8 - 9 | 1295 | 89 | ||
NM_001342985.1 | NP_001330723.1 | 8 - 9 | 1459 | 89 | ||
NM_001342986.1 | NP_001330724.1 | 8 - 9 | 1471 | 89 | ||
GenBank mRNA | AB493742.1 | - | 5 - 6 | 600 | 89 | |
BT029780.1 | - | 5 - 6 | 600 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |