Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127823.3 | NP_565539.1 | 2 - 3 | 417 | 102 | |
NM_201782.3 | NP_973511.1 | 2 - 3 | 417 | 102 | ||
GenBank mRNA | BT002359.1 | - | 2 - 3 | 331 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |