Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036226.2 | NP_001031303.1 | 10 - 11 | 1315 | 113 | |
NM_001198511.1 | NP_001185440.1 | 11 - 12 | 1216 | 113 | ||
NM_001334924.1 | NP_001323193.1 | 6 - 7 | 797 | 113 | ||
NM_106631.5 | NP_178100.3 | 11 - 12 | 1408 | 113 | ||
NM_180652.3 | NP_850983.1 | 11 - 12 | 1202 | 113 | ||
GenBank mRNA | AK317667.1 | - | 10 - 11 | 1129 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |