Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339736.1 | NP_001327282.1 | 3 - 4 | 569 | 112 | |
NM_001339737.1 | NP_001327281.1 | 1 - 2 | 668 | 112 | ||
NM_001339738.1 | NP_001327280.1 | 2 - 3 | 602 | 112 | ||
NM_001339739.1 | NP_001327279.1 | 3 - 4 | 331 | 112 | ||
NM_115421.4 | NP_567026.3 | 4 - 5 | 490 | 112 | ||
NM_180686.2 | NP_851017.1 | 4 - 5 | 431 | 112 | ||
NM_180687.2 | NP_851018.1 | 4 - 5 | 423 | 112 | ||
GenBank mRNA | AJ292545.1 | - | 4 - 5 | 321 | 112 | |
AY050867.1 | - | 4 - 5 | 395 | 112 | ||
AY091206.1 | - | 3 - 4 | 312 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |