Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203415.1 | NP_001190344.1 | 5 - 6 | 869 | 155 | |
NM_122011.4 | NP_568390.2 | 5 - 6 | 849 | 155 | ||
NM_180712.3 | NP_851043.1 | 5 - 6 | 849 | 155 | ||
GenBank mRNA | AB062615.1 | - | 5 - 6 | 659 | 155 | |
AF360208.1 | - | 5 - 6 | 774 | 155 | ||
AY040056.1 | - | 5 - 6 | 659 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |