Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPN20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343684.1 | NP_001318614.1 | 5 - 6 | 839 | 82 | |
NM_001343685.1 | NP_001331898.1 | 5 - 6 | 817 | 82 | ||
NM_122079.4 | NP_197572.1 | 5 - 6 | 669 | 82 | ||
NM_180714.4 | NP_851045.1 | 5 - 6 | 679 | 82 | ||
GenBank mRNA | AB007130.1 | - | 5 - 6 | 646 | 82 | |
AF428339.1 | - | 5 - 6 | 667 | 82 | ||
AF428366.1 | - | 5 - 6 | 663 | 82 | ||
AY034979.1 | - | 5 - 6 | 667 | 82 | ||
AY062971.1 | - | 4 - 5 | 586 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |