Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122080.3 | NP_568400.2 | 4 - 5 | 912 | 70 | |
NM_180715.1 | NP_851046.1 | 4 - 5 | 912 | 70 | ||
NM_180716.3 | NP_851047.1 | 4 - 5 | 978 | 70 | ||
GenBank mRNA | AY669789.1 | - | 4 - 5 | 303 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |