Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036852.4 | NP_001031929.4 | 8 - 9 | 1701 | 131 | |
NM_001343803.1 | NP_001331260.1 | 7 - 8 | 1540 | 131 | ||
NM_001343804.1 | NP_001331261.1 | 8 - 9 | 1522 | 131 | ||
NM_122252.4 | NP_568432.1 | 7 - 8 | 1694 | 131 | ||
NM_180734.3 | NP_851065.1 | 7 - 8 | 1560 | 131 | ||
GenBank mRNA | AB046717.1 | - | 6 - 7 | 1200 | 131 | |
AY139759.1 | - | 7 - 8 | 1616 | 131 | ||
BT004544.1 | - | 6 - 7 | 1200 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |