Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343982.1 | NP_001331385.1 | 4 - 5 | 511 | 92 | |
NM_122560.2 | NP_568488.2 | 4 - 5 | 453 | 92 | ||
NM_180749.4 | NP_851080.1 | 4 - 5 | 511 | 92 | ||
NM_180750.2 | NP_851081.1 | 4 - 5 | 453 | 92 | ||
GenBank mRNA | AK318672.1 | - | 4 - 5 | 445 | 92 | |
BT001097.1 | - | 4 - 5 | 349 | 92 | ||
BT001113.1 | - | 4 - 5 | 349 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |