Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036879.2 | NP_001031956.1 | 7 - 8 | 801 | 92 | |
NM_001036880.3 | NP_001031957.1 | 7 - 8 | 884 | 92 | ||
NM_001036881.1 | NP_001031958.1 | 4 - 5 | 621 | 92 | ||
NM_001085159.1 | NP_001078628.1 | 7 - 8 | 778 | 92 | ||
NM_122685.3 | NP_198154.1 | 7 - 8 | 927 | 92 | ||
NM_180756.3 | NP_851087.1 | 6 - 7 | 1111 | 92 | ||
GenBank mRNA | AB024283.1 | - | 6 - 7 | 1100 | 92 | |
AF424598.1 | - | 7 - 8 | 785 | 92 | ||
AY124831.1 | - | 6 - 7 | 742 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |