Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344092.1 | NP_001330486.1 | 2 - 3 | 501 | 101 | |
NM_122885.2 | NP_198344.2 | 4 - 5 | 829 | 101 | ||
NM_180761.1 | NP_851092.1 | 4 - 5 | 1006 | 101 | ||
NM_180762.3 | NP_851093.1 | 5 - 6 | 995 | 101 | ||
GenBank mRNA | AK229275.1 | - | 5 - 6 | 855 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |