Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125941.1 | NP_001119413.1 | 15 - 16 | 1576 | 71 | |
NM_124502.3 | NP_199936.2 | 16 - 17 | 1568 | 71 | ||
NM_180837.3 | NP_851168.1 | 16 - 17 | 1675 | 71 | ||
GenBank mRNA | AB053171.1 | - | 16 - 17 | 1520 | 71 | |
AY140086.1 | - | 16 - 17 | 1517 | 71 | ||
BT008388.1 | - | 15 - 16 | 1257 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |