Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125941.1 | NP_001119413.1 | 18 - 19 | 1933 | 96 | |
NM_124502.3 | NP_199936.2 | 19 - 20 | 1925 | 96 | ||
NM_180837.3 | NP_851168.1 | 19 - 20 | 2032 | 96 | ||
GenBank mRNA | AB053171.1 | - | 19 - 20 | 1877 | 96 | |
AY140086.1 | - | 19 - 20 | 1874 | 96 | ||
BT008388.1 | - | 18 - 19 | 1614 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |