Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345288.1 | NP_001318824.1 | 12 - 13 | 2253 | 75 | |
NM_001345289.1 | NP_001330355.1 | 11 - 12 | 1892 | 75 | ||
NM_001345290.1 | NP_001330356.1 | 10 - 11 | 2242 | 75 | ||
NM_125199.2 | NP_568874.2 | 11 - 12 | 1712 | 75 | ||
NM_180879.2 | NP_851210.1 | 11 - 12 | 2069 | 75 | ||
NM_180880.2 | NP_851211.1 | 11 - 12 | 2065 | 75 | ||
NM_180881.3 | NP_851212.2 | 10 - 11 | 1850 | 75 | ||
GenBank mRNA | AF053941.2 | - | 10 - 11 | 1607 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |