Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125495.2 | NP_568932.2 | 6 - 7 | 1396 | 105 | |
NM_180905.3 | NP_851236.1 | 6 - 7 | 1459 | 105 | ||
GenBank mRNA | AK226952.1 | - | 6 - 7 | 1402 | 105 | |
AY065413.1 | - | 6 - 7 | 1372 | 105 | ||
AY096551.1 | - | 6 - 7 | 1297 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |