Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125508.2 | NP_200923.2 | 1 - 2 | 314 | 77 | |
NM_180906.2 | NP_851237.1 | 1 - 2 | 314 | 77 | ||
GenBank mRNA | BT002058.1 | - | 1 - 2 | 223 | 77 | |
BT008386.1 | - | 1 - 2 | 117 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |