Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035762.6 | NP_001030839.5 | 3 - 4 | 2717 | 65 | |
NM_115036.7 | NP_190745.6 | 3 - 4 | 2711 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |