Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203135.1 | NP_001190064.1 | 5 - 6 | 533 | 96 | |
NM_115068.5 | NP_190777.1 | 4 - 5 | 420 | 96 | ||
GenBank mRNA | U28048.1 | - | 4 - 5 | 369 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |