Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115154.3 | NP_566975.1 | 12 - 13 | 1175 | 99 | |
NM_202691.1 | NP_974420.1 | 12 - 13 | 1134 | 99 | ||
GenBank mRNA | AF256535.1 | - | 12 - 13 | 1083 | 99 | |
AK227189.1 | - | 12 - 13 | 1041 | 99 | ||
AY064964.1 | - | 12 - 13 | 1143 | 99 | ||
AY133650.1 | - | 12 - 13 | 959 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |