Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115171.3 | NP_190879.1 | 7 - 8 | 1467 | 55 | |
GenBank mRNA | BT002444.1 | - | 7 - 8 | 1418 | 55 | |
BT008867.1 | - | 6 - 7 | 1336 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |