Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115208.4 | NP_190916.1 | 17 - 18 | 3022 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |