Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339642.1 | NP_001326036.1 | 2 - 3 | 294 | 130 | |
NM_115252.3 | NP_190960.1 | 2 - 3 | 294 | 130 | ||
GenBank mRNA | AF015544.1 | - | 2 - 3 | 143 | 130 | |
BT030362.1 | - | 2 - 3 | 134 | 130 | ||
D89994.1 | - | 2 - 3 | 263 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |