Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TON1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115357.4 | NP_567012.1 | 5 - 6 | 715 | 89 | |
GenBank mRNA | AK117295.1 | - | 5 - 6 | 641 | 89 | |
BT024487.1 | - | 5 - 6 | 563 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |