Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115587.2 | NP_191286.1 | 1 - 2 | 146 | 80 | |
GenBank mRNA | DQ446773.1 | - | 1 - 2 | 75 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |