Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339876.1 | NP_001326797.1 | 1 - 2 | 865 | 74 | |
NM_115613.4 | NP_191312.2 | 1 - 2 | 880 | 74 | ||
GenBank mRNA | BT011630.1 | - | 1 - 2 | 581 | 74 | |
BT012274.1 | - | 1 - 2 | 527 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |