Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339881.1 | NP_001325828.1 | 11 - 12 | 2140 | 122 | |
NM_115626.1 | NP_191325.1 | 11 - 12 | 1930 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |