Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAZ10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161241.2 | NP_001154713.1 | 2 - 3 | 453 | 125 | |
NM_001343260.1 | NP_001330992.1 | 2 - 3 | 495 | 125 | ||
NM_001343261.1 | NP_001330993.1 | 2 - 3 | 453 | 125 | ||
NM_121325.3 | NP_568287.1 | 2 - 3 | 495 | 125 | ||
NM_203046.3 | NP_974775.1 | 2 - 3 | 495 | 125 | ||
NM_203047.3 | NP_974776.1 | 2 - 3 | 495 | 125 | ||
GenBank mRNA | AY056444.1 | - | 2 - 3 | 364 | 125 | |
AY081715.1 | - | 2 - 3 | 280 | 125 | ||
FJ417331.1 | - | 2 - 3 | 280 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |