Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUVR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085233.2 | NP_001078702.1 | 9 - 10 | 2196 | 74 | |
NM_001085234.2 | NP_001078703.1 | 9 - 10 | 2283 | 74 | ||
NM_001203540.1 | NP_001190469.1 | 9 - 10 | 2319 | 74 | ||
NM_001344513.1 | NP_001331768.1 | 9 - 10 | 2297 | 74 | ||
NM_001344514.1 | NP_001331767.1 | 9 - 10 | 2297 | 74 | ||
NM_001344515.1 | NP_001331773.1 | 8 - 9 | 2355 | 74 | ||
NM_001344516.1 | NP_001331766.1 | 8 - 9 | 2281 | 74 | ||
NM_001344517.1 | NP_001331772.1 | 9 - 10 | 2221 | 74 | ||
NM_001344518.1 | NP_001331771.1 | 9 - 10 | 2207 | 74 | ||
NM_001344519.1 | NP_001331770.1 | 9 - 10 | 2207 | 74 | ||
NM_001344520.1 | NP_001331769.1 | 9 - 10 | 2133 | 74 | ||
NM_001344521.1 | NP_001331765.1 | 8 - 9 | 2445 | 74 | ||
NM_001344522.1 | NP_001331764.1 | 8 - 9 | 2385 | 74 | ||
NM_001344523.1 | NP_001331763.1 | 8 - 9 | 2371 | 74 | ||
NM_001344524.1 | NP_001331762.1 | 9 - 10 | 2357 | 74 | ||
NM_123766.3 | NP_568631.1 | 9 - 10 | 2209 | 74 | ||
NM_203151.3 | NP_974880.1 | 9 - 10 | 2250 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |