Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125287.5 | NP_568894.1 | 4 - 5 | 790 | 81 | |
NM_203233.1 | NP_974962.1 | 4 - 5 | 756 | 81 | ||
GenBank mRNA | AB021706.1 | - | 4 - 5 | 531 | 81 | |
JQ937241.1 | - | 4 - 5 | 516 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |