Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198200.2 | NP_001185129.1 | 17 - 18 | 3355 | 76 | |
NM_001333042.1 | NP_001319133.1 | 17 - 18 | 3343 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |