Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333084.1 | NP_001322412.1 | 9 - 10 | 709 | 92 | |
NM_103129.5 | NP_564436.4 | 9 - 10 | 730 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |