Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103364.3 | NP_174901.1 | 2 - 3 | 1353 | 86 | |
GenBank mRNA | AF367272.1 | - | 2 - 3 | 1272 | 86 | |
AF436835.1 | - | 2 - 3 | 1278 | 86 | ||
AK230230.1 | - | 2289 | 86 | |||
AY037183.1 | - | 2 - 3 | 1278 | 86 | ||
AY039914.1 | - | 2 - 3 | 1272 | 86 | ||
AY133530.1 | - | 2 - 3 | 1188 | 86 | ||
AY142568.1 | - | 2 - 3 | 1188 | 86 | ||
J03240.1 | - | 2 - 3 | 1266 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |