Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103456.4 | NP_174996.1 | 4 - 5 | 729 | 60 | |
GenBank mRNA | AK230314.1 | - | 3439 | 60 | ||
AY039539.2 | - | 4 - 5 | 601 | 60 | ||
AY039961.1 | - | 4 - 5 | 649 | 60 | ||
AY079402.1 | - | 4 - 5 | 530 | 60 | ||
AY095991.1 | - | 4 - 5 | 595 | 60 | ||
AY140091.1 | - | 4 - 5 | 570 | 60 | ||
BT002267.1 | - | 4 - 5 | 530 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |