Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103536.4 | NP_175076.2 | 4 - 5 | 1253 | 67 | |
GenBank mRNA | BT002389.1 | - | 4 - 5 | 1100 | 67 | |
BT009680.1 | - | 4 - 5 | 791 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |