Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103546.4 | NP_175086.1 | 2 - 3 | 957 | 68 | |
GenBank mRNA | AK229399.1 | - | 2 - 3 | 682 | 68 | |
AY072322.1 | - | 2 - 3 | 681 | 68 | ||
AY074380.1 | - | 2 - 3 | 616 | 68 | ||
AY117292.1 | - | 2 - 3 | 596 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |