Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCA3;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103617.4 | NP_175156.1 | 1 - 2 | 499 | 125 | |
NM_202254.2 | NP_973983.1 | 1 - 2 | 496 | 125 | ||
GenBank mRNA | AK226253.1 | - | 1 - 2 | 502 | 125 | |
AY087628.1 | - | 1 - 2 | 481 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |