Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333320.1 | NP_001321802.1 | 7 - 8 | 986 | 77 | |
NM_103724.2 | NP_175261.1 | 7 - 8 | 986 | 77 | ||
GenBank mRNA | AK228479.1 | - | 7 - 8 | 839 | 77 | |
U95143.1 | - | 7 - 8 | 982 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |