Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203206.1 | NP_001190135.1 | 12 - 13 | 1926 | 73 | |
NM_001339969.1 | NP_001326041.1 | 12 - 13 | 2793 | 73 | ||
NM_001339970.1 | NP_001326042.1 | 11 - 12 | 1935 | 73 | ||
NM_001339971.1 | NP_001326040.1 | 11 - 12 | 1706 | 73 | ||
NM_115803.3 | NP_191500.2 | 11 - 12 | 1825 | 73 | ||
GenBank mRNA | AJ459823.1 | - | 11 - 12 | 1473 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |