Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115866.4 | NP_191562.1 | 3 - 4 | 1766 | 69 | |
GenBank mRNA | AB493658.1 | - | 3 - 4 | 1192 | 69 | |
AJ132096.1 | - | 3 - 4 | 1192 | 69 | ||
BT003817.1 | - | 3 - 4 | 1192 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |