Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115927.5 | NP_191622.1 | 1 | 2192 | 101 | |
GenBank mRNA | AY064062.1 | - | 1 | 2038 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |