Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP65-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340060.1 | NP_001319808.1 | 8 - 9 | 1337 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |