Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336238.1 | NP_001324058.1 | 4 - 5 | 1124 | 84 | |
NM_128562.4 | NP_180568.1 | 4 - 5 | 819 | 84 | ||
GenBank mRNA | AF029876.1 | - | 4 - 5 | 590 | 84 | |
AF033118.1 | - | 4 - 5 | 624 | 84 | ||
AK221387.1 | - | 4 - 5 | 686 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |